1-Ethynylcyclohexanol ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1-Ethynyl-1-cyclohexanol
CAS Number:78-27-3
:
Formula:C₈H₁₂O
MDL Number:MFCD00003858
:124.18 g/mol
Boiling Pt:180 °C
Melting Pt:30…33 °C
Density:0,973
Flash Pt:73 °C (163 °F)
Storage Temperature:Ambient
EINECS:201-100-9
UN:2811
ADR:6.1,III
:GHS06
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together