Order Entry
Ireland
ContactUsLinkComponent
Hydrochloric acid 6 mol/l (6 N)
Irritant
Hydrochloric acid 6 mol/l (6 N)
Catalog # VWRUVW3204-1
Supplier:  VWR Collection
CAS Number:  
Irritant
Hydrochloric acid 6 mol/l (6 N)
Catalog # VWRUVW3204-1
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
Density:1,06 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
ADR:8,II
 :36.46 g/mol
UN:1789
 :GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324