Order Entry
Ireland
ContactUsLinkComponent
Supelco® Organic Reference Standard, (+)-Menthone
Supelco® Organic Reference Standard, (+)-Menthone
Catalog # 63675-1ML-F
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, (+)-Menthone
Catalog # 63675-1ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    3391-87-5
  • Description:
    (+)-Menthone
  • Storage conditions:
    2…8 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00210063
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (1S,4R)-p-Menthan-3-one; (2R,5S)-2-Isopropyl-5-methylcyclohexanone
  • Molecular weight:
    154,25
  • Density:
    0,893 g/ml at 20 °C (lit.)
  • Flash point:
    72 °C - Closed cup
  • Boiling point:
    209 - 210 °C (lit.)

Specifications

About this item