4-Picolyl chloride hydrochloride
Catalog # MOLEM44972750
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
About this item
Synonyms: 4-(Chloromethyl)pyridine hydrochloride, 4-(Chloromethyl)-pyridinium chloride
:
ADR:8,III
:164.03 g/mol
EINECS:217-350-7
UN:3261
CAS Number:1822-51-1
Formula:C₆H₇Cl₂N
MDL Number:MFCD00012826