2,4,6-Triphenylpyrilium tetrafluoroborate, Sigma-Aldrich®
Catalog # 8.18238.0005
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Melting Point:252 - 255 °C
- Molar Mass:396.2 g/mol
- Hill Formula:C23H17BF4O
Specification Test Results
About this item
:
:396.19 g/mol
:M-Clarity™ quality level = MQ200
EINECS:207-186-4
CAS Number:448-61-3
Formula:C₂₃H₁₇BF₄O
MDL Number:MFCD00012001