Organic reference standard, Dihexyl phthalate 100 µg/ml in n-nonane, CIL
Catalog # PROOCILULM74341.2
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:84-75-3
- Description:Dihexyl phthalate 100 µg/ml in n-nonane
- Formula:C₂₀H₃₀O₄
- Description:Dihexyl phthalate