
3-Formyl-2-methoxypyridine ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:71255-09-9
:
Formula:C₇H₇NO₂
MDL Number:MFCD04115112
:137.14 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together