



Cyclohexane, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-82-7
:
Formula:C₆H₁₂
MDL Number:MFCD00003814
:84.16 g/mol
Merck Index:13,02752
Boiling Pt:81 °C (760 mmHg)
Melting Pt:6,5 °C
Density:0,77 g/cm³
Flash Pt:–18 °C
EINECS:203-806-2
UN:1145
ADR:3,II
:GHS07
:GHS08
:GHS09
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 % (Coulometric)
- GC:≥99.4 %
- Acidity:≤0.001 meq/g
Specification Test Results
Related Information
Frequently Bought Together