2,4,6-Tri-tert-butylaniline 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:961-38-6
:
Formula:C₁₈H₃₁N
MDL Number:MFCD00011645
:261.45 g/mol
Boiling Pt:302 °C (1013 hPa)
Melting Pt:144…147 °C
Density:0,991 g/cm³ (19.85 °C)
Storage Temperature:Ambient
EINECS:213-507-9
UN:2811
ADR:6.1,III
:GHS06
:GHS07
Related Information
Frequently Bought Together