2,4,6-Trimethylbenzaldehyde 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:487-68-3
:
Formula:C₁₀H₁₂O
MDL Number:MFCD00003341
:148.2 g/mol
Boiling Pt:237 °C (1013 hPa)
Melting Pt:14 °C
Density:1 g/cm³
Flash Pt:105 °C
EINECS:207-662-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5520 to 1.5540 (20°C, 589 nm)
- GC:≥96.0 %
- Loss on ignition:24.5 % to 26.5 % (800°C)
Specification Test Results
Frequently Bought Together