Order Entry
Ireland
ContactUsLinkComponent
p-Xylene reference substance for gas chromatography, Supelco®
Flammable
Irritant
Health
p-Xylene reference substance for gas chromatography, Supelco®
Catalog # 1.09799.0005
Supplier:  Merck
CAS Number:  
undefined
Flammable
Irritant
Health
p-Xylene reference substance for gas chromatography, Supelco®
Catalog # 1.09799.0005
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Solubility:
    0.2 g/l (25 °C)
  • Melting Point:
    13.3 °C
  • Molar Mass:
    106.17 g/mol
  • Boiling Point:
    138.4 °C (1013 hPa)
  • Vapor Pressure:
    8.7 hPa (20 °C)
  • Flash Point:
    27.2 °C
  • Explosion Limit:
    1.1 - 7 %(V)
  • Density:
    0.86 g/cm3 (20 °C)
  • Ignition Point:
    540 °C
  • Saturation concentration:
    38 g/m3 (20 °C) Air
  • Evaporation number:
    13.5
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable liquid

Specification Test Results

About this item

 :
 :106.17 g/mol
Merck Index:12,10214
Boiling Pt:138,3 °C (1013 hPa)
Melting Pt:13 °C
Density:1,096 g/cm³ (20 °C)
Flash Pt:25 °C
Storage Temperature:Ambient
EINECS:203-396-5
UN:1307
ADR:3,III
 :M-Clarity™ quality level = MQ100
 :GHS02
 :GHS07
 :GHS08
CAS Number:106-42-3
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00008556