
2,5-Dimethylbenzoic acid 98%
:610-72-0
:
:C₉H₁₀O₂
:MFCD00002482
:150.18 g/mol
:268 °C (760 mmHg)
:131…135 °C
:1,06 g/cm³ (20 °C)
:Ambient
:210-235-2
:GHS07
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:131 to 135 °C
- Titration with NaOH:≥97.5 %
Frequently Bought Together