Order Entry
Ireland
ContactUsLinkComponent
(R)-(+)-4-Methoxy-α-methylbenzylamine ≥99%, ee 99%, ChiPros®
Corrosive
Irritant
(R)-(+)-4-Methoxy-α-methylbenzylamine ≥99%, ee 99%, ChiPros®
Catalog # L16321.03
CAS Number:  
Corrosive
Irritant
(R)-(+)-4-Methoxy-α-methylbenzylamine ≥99%, ee 99%, ChiPros®
Catalog # L16321.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:22038-86-4
 :
Formula:C₉H₁₃NO
Boiling Pt:65 °C (0,38 mmHg)
Melting Pt:< –20 °C
Density:1,02
UN:2735
ADR:8,III
 :151.21 g/mol
 :GHS05
 :GHS07
Storage Temperature:Ambient
MDL Number:MFCD00671659