Order Entry
Ireland
ContactUsLinkComponent
4-Morpholinoaniline 98+%
Irritant
4-Morpholinoaniline 98+%
Certificates
undefined
Irritant
4-Morpholinoaniline 98+%

Specification Test Results

  • Appearance:
    Brown to purple Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    128 to 132 °C
  • Titration with HClO4:
    ≥98.0 %

Specification Test Results

Related Information

Frequently Bought Together