Order Entry
Ireland
ContactUsLinkComponent
 
1,3-Dicyclohexylurea 98%
 
undefined
1,3-Dicyclohexylurea 98%

 

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    231 to 236 °C
  • Titration with HClO4:
    ≥97.5 %
  • HPLC:
    ≥99.0 % (ELSD)
  • Dnase,Rnase,Protease:
    None detected
  • Heavy metals (as Pb):
    ≤5 ppm

 

 

Frequently Bought Together