
4-tert-Butyl-1-(bromomethyl)benzene 97%
:18880-00-7
:
:C₁₁H₁₅Br
:MFCD00000180
:227.14 g/mol
:90…94 °C (1,5 mmHg)
:8…12 °C
:1,236 g/cm³
:>109 °C
:Ambient
:242-643-1
:3265
:8,II
:GHS05
- Appearance:Clear light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5445 to 1.5465 (20°C, 589 nm)
- Titration Argentometric:≥96.0 %
Frequently Bought Together