Order Entry
Ireland
ContactUsLinkComponent
Stopcocks for burettes, angled, DURAN®
Stopcocks for burettes, angled, DURAN®
Catalog # SCOT286451608
Supplier:  DWK Life Sciences
Stopcocks for burettes, angled, DURAN®
Catalog # SCOT286451608
Supplier:  DWK Life Sciences
Specifications
  • Hole Ø:
    2,0 mm
  • Environmentally Preferable:
  • Pk:
    50
  • Spigot:
    12,5 NS
Specifications
About this item
DURAN®, borosilicate glass 3.3, clear, grounded and polished, without key.
  • Very good chemical resistance
  • Very good temperature resistance
For glass manipulators.