Order Entry
Ireland
ContactUsLinkComponent
Toluene ≥99.0%, GPR RECTAPUR®
Toluene ≥99.0%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Toluene ≥99.0%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Appearance:
    Clear colourless liquid
  • IR Spectrum:
    Passes test
  • Boiling point:
    109 - 112 °C
  • Density (20/4):
    0.865 - 0.868
  • Benzene:
    Max. 0.02 %
  • Evaporation residue:
    Max. 50 ppm
  • Water:
    Max. 0.1 %

Specification Test Results

Frequently Bought Together