Tubing connectors, straight, conical nozzles
Supplier: BURKLE
Certificates
About this item
Made from PP, white.
- Sterilisable
- Autoclavable up to max. 121 °C (20 min.)
- Suitable for use with foodstuffs
- Good chemical resistance
Conical, straight hose connectors for various hose sizes.