


Toluene ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:108-88-3
:
Formula:C₆H₅CH₃
MDL Number:MFCD00008512
:92.14 g/mol
Merck Index:14,9529
Boiling Pt:111 °C
Melting Pt:–93 °C
Density:0,867
Flash Pt:4 °C (40 °F)
Storage Temperature:Ambient
EINECS:203-625-9
UN:1294
ADR:3,II
REACH:05-2114615130-69
:GHS02
:GHS08
:GHS07
Specification Test Results
- Form:Liquid
- Color (APHA):≤10
- Assay (GC):>99.5%
- Residual water:≤0.03%
- Impurity content::Substances darkened by sulphuric acid P.T.
- Residue on evaporation:≤0.001%
Specification Test Results
Related Information
Frequently Bought Together