Order Entry
Ireland
ContactUsLinkComponent
Soap and disinfectant dispenser
Soap and disinfectant dispenser
Catalog # HUPF8600129
Supplier:  HUPFER METALLWERKE
undefined
Soap and disinfectant dispenser
Catalog # HUPF8600129
Supplier:  HUPFER METALLWERKE

Specifications

  • Capacity:
    1000 ml
  • Colour:
    Aluminium
  • Description:
    Lotion soap dispenser
  • W×D×H:
    93×197×302 mm
  • Pk:
    1

Specifications

About this item

This dispenser made from stainless steel can be either filled with liquid soaps, washing lotions, and disinfectants.

  • Arm lever for improved hygiene
  • Refillable plastic reservoir with large filling opening
  • Filling level visible to the user for easier maintenance
  • High quality plastic pump with adjustable dosage amount

Colour: Aluminium.