

Benzylamine ≥99.0%, LiChropur™ for GC derivatisation, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:100-46-9
:
Formula:(C₆H₅)CH₂NH₂
MDL Number:MFCD00008106
:107.16 g/mol
Merck Index:13,01127
Boiling Pt:185 °C (1013 hPa)
Melting Pt:10 °C
Density:0,98 g/cm³ (20 °C)
EINECS:202-854-1
UN:2735
ADR:8,II
:M-Clarity™ quality level = MQ100
Flash Pt:65 °C
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together