Jars, wide mouth, round, with screw cap
: BURKLE
HDPE, translucent.
- Good chemical resistance
- Good temperature resistance
- Screw lid made of PP with seal made of PE
Wide opening makes it ideally suited for storing or shipping powders and granules.
Frequently Bought Together