Order Entry
Ireland
ContactUsLinkComponent
α-Bromo-3-fluorotoulene 95%
α-Bromo-3-fluorotoulene 95%
Certificates
undefined
α-Bromo-3-fluorotoulene 95%

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5450 to 1.5480 (20°C, 589 nm)
  • GC:
    ≥94 %

Specification Test Results

Related Information

Frequently Bought Together