
Pentafluorobenzyl bromide
Catalog # MOLE37860017-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:260.99 g/mol
Boiling Pt:174 °C (1013 hPa)
Density:1,72 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:217-182-4
UN:3265
ADR:8,II
:GHS07
CAS Number:1765-40-8
Formula:(C₆F₅)CH₂Br
MDL Number:MFCD00000299