Pentafluorobenzyl bromide for synthesis, Sigma-Aldrich®
Catalog # 8.41643.0005
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 ml
- Seq:2210
Specifications
Specification Test Results
- Molar Mass:260.99 g/mol
- Boiling Point:174 - 175 °C
- Flash Point:83 °C
- Density:1.85 g/cm3 (20 °C)
Specification Test Results
About this item
:
:260.99 g/mol
Boiling Pt:174 °C (1013 hPa)
Density:1,72 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:6.1,II
:M-Clarity™ quality level = MQ200
EINECS:217-182-4
UN:2810
:GHS07
CAS Number:1765-40-8
Formula:(C₆F₅)CH₂Br
MDL Number:MFCD00000299