
3-Bromobenzyl chloride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:932-77-4
:
Formula:C₇H₆BrCl
MDL Number:MFCD00040865
:205.48 g/mol
Boiling Pt:118…119 °C (18 mmHg)
Melting Pt:32,5 °C
Density:1,608
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Related Information
Frequently Bought Together