Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, 1,3-Dinitrobenzene 1000 µg/ml in acetonitrile, CIL
Organic reference standard, 1,3-Dinitrobenzene 1000 µg/ml in acetonitrile, CIL
  PROOCILULM38501.2
 :  
Organic reference standard, 1,3-Dinitrobenzene 1000 µg/ml in acetonitrile, CIL
  PROOCILULM38501.2
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    99-65-0
  • Description:
    1,3-Dinitrobenzene 1,000 µg/ml in acetonitrile
  • Formula:
    C₆H₄N₂O₄
  • Description:
    1,3-Dinitrobenzene