Order Entry
Ireland
ContactUsLinkComponent
Phenol ≥99%, detached crystals, GPR RECTAPUR®
Toxic
Corrosive
Health
Phenol ≥99%, detached crystals, GPR RECTAPUR®
Catalog # 20598.465
Supplier:  VWR Chemicals
CAS Number:  
undefined
Toxic
Corrosive
Health
Phenol ≥99%, detached crystals, GPR RECTAPUR®
Catalog # 20598.465
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Metal drum
  • Pk:
    25 kg
  • Seq:
    4283

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Congealing temperature:
    Min. 39.5 °C
  • Evaporation residue:
    Max. 0.025 %

Specification Test Results

About this item

 :
 :94.11 g/mol
Boiling Pt:181,8 °C (1013 hPa)
Melting Pt:40,8 °C
Density:1,06 g/cm³ (20 °C)
Flash Pt:81 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-632-7
UN:1671
ADR:6.1,II
REACH:01-2119471329-32
 :GHS06
 :GHS05
 :GHS08
CAS Number:108-95-2
Formula:C₆H₅OH
MDL Number:MFCD00002143