


Phenol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:108-95-2
:
Formula:C₆H₅OH
MDL Number:MFCD00002143
:94.11 g/mol
Merck Index:12,07390
Boiling Pt:181,7 °C (1013 hPa)
Melting Pt:40,8 °C
Density:1,0722 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-632-7
UN:1671
ADR:6.1,II
:M-Clarity™ quality level = MQ200
Flash Pt:81 °C
REACH:01-2119471329-32
:GHS06
:GHS05
:GHS08
Specification Test Results
- Solubility:84 g/l (20 °C)
- Melting Point:40.8 °C
- Molar Mass:94.11 g/mol
- Bulk Density:620 kg/m3
- Boiling Point:181.8 °C (1013 hPa)
- Vapor Pressure:0.2 hPa (20 °C)
- Flash Point:81 °C
- Explosion Limit:1.3 - 9.5 %(V)
- Density:1.06 g/cm3 (20 °C)
- pH:5 (50 g/l, H2O, 20 °C)
- Ignition Point:595 °C
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Specification Test Results
Related Information
Frequently Bought Together