Order Entry
Ireland
ContactUsLinkComponent
Phenol, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Toxic
Corrosive
Health
Phenol, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
Toxic
Corrosive
Health
Phenol, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay (by GC):
    min. 99.0 %
  • Clarity of Solution:
    passes test
  • Freezing Point:
    min. 40.5 °C
  • Residue after Evaporation:
    max. 0.03 %
  • Water (H2O):
    max. 0.2 %

Specification Test Results

Related Information

Frequently Bought Together