Order Entry
Ireland
ContactUsLinkComponent
1-Boc-6-methoxyindole-2-boronic acid ≥98%
Irritant
1-Boc-6-methoxyindole-2-boronic acid ≥98%
Catalog # H52482.MD
CAS Number:  
Irritant
1-Boc-6-methoxyindole-2-boronic acid ≥98%
Catalog # H52482.MD
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

CAS Number:850568-65-9
 :
Formula:C₁₄H₁₈BNO₅
MDL Number:MFCD06659830
 :291.11 g/mol
Melting Pt:100…103 °C
 :GHS07