
Ethyl-4-bromobutyrate 97+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2969-81-5
:
Formula:C₆H₁₁BrO₂
MDL Number:MFCD00000259
:195.06 g/mol
Boiling Pt:80…82 °C (10 mmHg)
Density:1,36 g/cm³
Flash Pt:90 °C
EINECS:221-005-6
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4550 to 1.4570 (20°C, 589 nm)
- GC:≥97.0 %
Specification Test Results
Frequently Bought Together