Potassium hydroxide hydrate ≥99.995% (metals basis), TraceSELECT™ for trace analysis, Fluka™
Catalog # 60371-25G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Aluminium (Al):Max. 0.05 ppm
- Arsenic (As):Max. 0.05 ppm
- Barium (Ba):Max. 1 ppm
- Calcium (Ca):Max. 0.5 ppm
- Cadmium (Cd):Max. 0.01 ppm
- Cerium (Ce):Max. 0.01 ppm
- Cobalt (Co):Max. 0.01 ppm
- Chromium (Cr):Max. 0.01 ppm
- Copper (Cu):Max. 0.01 ppm
- Europium (Eu):Max. 0.01 ppm
- Iron (Fe):Max. 0.05 ppm
- Mercury (Hg):Max. 1 ppb
- Iridium (Ir):Max. 0.5 ppm
- Lanthanum (La):Max. 0.01 ppm
- Lithium (Li):Max. 0.5 ppm
- Magnesium (Mg):Max. 0.1 ppm
- Manganese (Mn):Max. 0.05 ppm
- Sodium (Na):Max. 5 ppm
- Nickel (Ni):Max. 0.01 ppm
- Lead (Pb):Max. 0.01 ppm
- Palladium (Pd):Max. 0.5 ppm
- Platinum (Pt):Max. 0.5 ppm
- Rhodium (Rh):Max. 0.5 ppm
- Ruthenium (Ru):Max. 0.5 ppm
- Scandium (Sc):Max. 0.01 ppm
- Samarium (Sm):Max. 0.01 ppm
- Strontium (Sr):Max. 0.5 ppm
- Thallium (Tl):Max. 0.01 ppm
- Vanadium (V):Max. 0.1 ppm
- Yttrium (Y):Max. 0.01 ppm
- Ytterbium (Yb):Max. 0.01 ppm
- Zinc (Zn):Max. 0.05 ppm
- Silicate (as SiO₂):Max. 5 ppm
- Chloride (Cl):Max. 1000 ppb
- Phosphate (PO₄):Max. 50 ppb
- Sulfate (SO₄):Max. 5000 ppb
- Carbonate (K₂CO₃):Max. 0.6 %
- Total nitrogen (as N):Max. 5 ppm
Specification Test Results
About this item
:
:74.12 g/mol
Merck Index:12,07806
Boiling Pt:1,320 °C (1.013 hPa)
Melting Pt:143 °C
Density:2,04 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-181-3
UN:1813
ADR:8,II
REACH:01-2119487136-33
:GHS07
:GHS05
CAS Number:26288-25-5
Formula:H₃KO₂
MDL Number:MFCD00003553