Order Entry
Ireland
ContactUsLinkComponent
N-Acetyl-DL-tryptophan
N-Acetyl-DL-tryptophan
Catalog # PROOMM0172.13
CAS Number:  
N-Acetyl-DL-tryptophan
Catalog # PROOMM0172.13
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :246.27 g/mol
Melting Pt:205…206 °C
Storage Temperature:Ambient
EINECS:201-739-3
CAS Number:87-32-1
Formula:C₁₃H₁₄N₂O₃
MDL Number:MFCD00005644