Order Entry
Ireland
ContactUsLinkComponent
Hydroquinone monomethyl ether for synthesis, Sigma-Aldrich®
Hydroquinone monomethyl ether for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Hydroquinone monomethyl ether for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    40 g/l (25 °C)
  • Melting Point:
    54 - 57 °C
  • Molar Mass:
    124.14 g/mol
  • Boiling Point:
    243 °C (1013 hPa)
  • Vapor Pressure:
    0.13 hPa (60 °C)
  • Flash Point:
    133 °C
  • Density:
    1.25 g/cm3 (20 °C)
  • pH:
    5.1 (30 g/l, H2O, 20 °C)
  • Ignition Point:
    445 °C

Specification Test Results

Related Information

Frequently Bought Together