Order Entry
Ireland
Orders LinkContactUsLinkComponent
Hydroquinone monomethyl ether ≥98%

Hydroquinone monomethyl ether ≥98%

Hydroquinone monomethyl ether ≥98%
Certificates
Hydroquinone monomethyl ether ≥98%
Hydroquinone monomethyl ether ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98+% CAS No.: 150-76-5 Molecular Formula: C7H8O2 Formula Weight: 124.14 Flash Point: 132°(269°F) Melting Point: 55-58° Boiling Point: 243-246° Beilstein Reference: 507924 MDL No.: MFCD00002332

Synonyms:  p-Methoxyphenol, Mequinol, 4-Methoxyphenol, 1-Hydroxy-4-methoxybenzene, 4-Hydroxyanisole
  • CAS Number:150-76-5
  • Formula:C₇H₈O₂
  • MDL Number:MFCD00002332
  • Molecular Weight:124.14 g/mol
  • Boiling Pt:243…246 °C
  • Melting Pt:55…58 °C
  • Density:1,550
  • Flash Pt:132 °C (269 °F)
  • Storage Temperature:Ambient
  • EINECS:205-769-8
Product Family Options

Product Information