3,4,5-Trimethoxyaniline
Catalog # MOLEM33415626
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
About this item
:
:183.21 g/mol
EINECS:246-154-4
UN:2811
:GHS07
Storage Temperature:Ambient
CAS Number:24313-88-0
Formula:C₉H₁₃NO₃
MDL Number:MFCD00008393
Boiling Pt:293
Melting Pt:110…113 °C
ADR:6.1 III