Order Entry
Ireland
ContactUsLinkComponent
2,4-Dinitroaniline
Toxic
Health
Environment
2,4-Dinitroaniline
Catalog # MOLE56996925-500G
Supplier:  Molekula
CAS Number:  
Toxic
Health
Environment
2,4-Dinitroaniline
Catalog # MOLE56996925-500G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :183.12 g/mol
Merck Index:13,03298
Boiling Pt:401 °C (1013 hPa)
Melting Pt:177…178 °C
Density:1,6 g/cm³ (20 °C)
Flash Pt:224 °C
EINECS:202-553-5
UN:1596
ADR:6.1,II
 :GHS06
 :GHS08
 :GHS09
Storage Temperature:Ambient
CAS Number:97-02-9
Formula:C₆H₅N₃O₄
MDL Number:MFCD00007151