



Phenyl isocyanate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:103-71-9
:
Formula:C₇H₅NO
MDL Number:MFCD00001994
:119.12 g/mol
Merck Index:12,07449
Boiling Pt:165 °C (1013 hPa)
Melting Pt:–33 °C
Density:1,1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-137-6
UN:2487
ADR:6.1 (3),I
:M-Clarity™ quality level = MQ200
Flash Pt:51 °C
:GHS02
:GHS06
:GHS05
:GHS08
Frequently Bought Together