Methyl orange
VWRS1460.1000
:
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
:
:327.34 g/mol
:13,06127
:∼90 °C (1013 hPa)
:> 300 °C
:∼1 g/cm³ (20 °C)
:38 °C
:Ambient
:208-925-3
:3143
:6.1,III
:GHS06
:547-58-0
:C₁₄H₁₄N₃NaO₃S
:MFCD00007502