
Iodobenzene for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:591-50-4
:
Formula:C₆H₅I
MDL Number:MFCD00001029
Merck Index:13,05051
Boiling Pt:188 °C (1013 hPa)
Melting Pt:–30 °C
Density:1,838 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:74 °C
:204.01 g/mol
EINECS:209-719-6
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:0.34 g/l (30 °C) (experimental)
- Melting Point:-30 °C
- Molar Mass:204.01 g/mol
- Boiling Point:187 - 189 °C
- Vapor Pressure:1.41 hPa (25 °C)
- Flash Point:77 °C
- Refractive Index:1.6198 (20 °C, 589 nm)
- Density:1.83 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together