2,4,6-Trichlorophenol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:88-06-2
:
Formula:C₆H₃Cl₃O
MDL Number:MFCD00002172
:197.45 g/mol
Merck Index:14,9644
Boiling Pt:245…246 °C
Melting Pt:63…68 °C
Density:1,675
Flash Pt:99 °C (210 °F)
Storage Temperature:Ambient
EINECS:201-795-9
UN:2020
ADR:6.1,III
:GHS08
:GHS07
:GHS09
Related Information
Frequently Bought Together