2,3,4,5-Tetrachloroaniline
Catalog # MAYBBTB15037.1
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:230.91 g/mol
EINECS:211-216-1
UN:1648
ADR:3,II
CAS Number:634-83-3
Formula:C₆H₃Cl₄N