Order Entry
Ireland
ContactUsLinkComponent
Chlorobenzene for synthesis, Sigma-Aldrich®
Flammable
Irritant
Environment
Chlorobenzene for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Flammable
Irritant
Environment
Chlorobenzene for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    0.4 g/l (20 °C)
  • Melting Point:
    -45 °C
  • Molar Mass:
    112.56 g/mol
  • Boiling Point:
    132 °C (1013 hPa)
  • Vapor Pressure:
    12 hPa (20 °C)
  • Flash Point:
    27 °C
  • Refractive Index:
    1.5248 (20 °C, 589 nm)
  • Explosion Limit:
    1.3 - 11 %(V)
  • Density:
    1.11 g/cm3 (20 °C)
  • pH:
    (H2O, 20 °C) neutral
  • Saturation concentration:
    54 g/m3 (20 °C) Air
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable liquid

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together