Order Entry
Ireland
ContactUsLinkComponent
 
 
 
 
Chlorobenzene ≥99.5%, AnalaR NORMAPUR® analytical reagent
  103386D
 :  
undefined
 
 
 
Chlorobenzene ≥99.5%, AnalaR NORMAPUR® analytical reagent
  103386D
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

 

 

  • Assay:
    Min. 99.5 %
  • IR Spectrum:
    Passes test
  • Colouration:
    Max. 10 APHA
  • Density (20/4):
    1.100 - 1.110
  • Evaporation residue:
    Max. 0.05 %
  • Water:
    Max. 0.05 %
  • Al (Aluminium):
    Max. 2.0 ppm
  • Ba (Barium):
    Max. 1.0 ppm
  • Ca (Calcium):
    Max. 5.0 ppm
  • Cd (Cadmium):
    Max. 0.5 ppm
  • Co (Cobalt):
    Max. 0.2 ppm
  • Cr (Chromium):
    Max. 0.2 ppm
  • Cu (Copper):
    Max. 0.2 ppm
  • Fe (Iron):
    Max. 2.0 ppm
  • K (Potassium):
    Max. 1.0 ppm
  • Mg (Magnesium):
    Max. 0.5 ppm
  • Mn (Manganese):
    Max. 0.2 ppm
  • Na (Sodium):
    Max. 5.0 ppm
  • Ni (Nickel):
    Max. 0.2 ppm
  • Pb (Lead):
    Max. 0.5 ppm
  • Sr (Strontium):
    Max. 0.2 ppm
  • Zn (Zinc):
    Max. 1 ppm

 

 

 :
 :112.56 g/mol
 :13,02139
 :130 °C (1013 hPa)
 :–45 °C
 :1,11 g/cm³ (20 °C)
 :28 °C
 :Ambient
 :203-628-5
 :1134
 :3,III
 :01-2119432722-45
 :GHS02
 :GHS07
 :GHS09
 :108-90-7
 :C₆H₅Cl
 :MFCD00000530