Order Entry
Ireland
ContactUsLinkComponent
Trimesic acid 98%
Irritant
Trimesic acid 98%
Certificates
undefined
Irritant
Trimesic acid 98%
Certificates

Specification Test Results

  • Appearance:
    White to beige Powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥97.5 %
  • UV Lambda max:
    590 to 596 nm (in Methanol)
  • Molar absorption coefficient:
    67400 l/(mol.cm) (at Lambda max)

Specification Test Results

Related Information

Frequently Bought Together