


Salicylic acid 99.5-105.0% (by acidimetry on anhydrous substance), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
:69-72-7
:
:(HO)C₆H₄CO₂H
:MFCD00002439
:138.12 g/mol
:13,08411
:336 °C (1013 hPa)
:157…159 °C
:1,443 g/cm³ (20 °C)
:157 °C
:200-712-3
:GHS05
:GHS08
:GHS07
:Ambient
- Assay (acidimetric; calculated on dried substance):99.5 - 100.5 %
- Assay (HPLC, calc.on dried substance):98.0 - 102.0 %
- Identity:passes test
- Appearance of solution (10 %; ethanol 96 %):passes test
- Chloride (Cl):≤ 0.0100 %
- Sulfate (SO₄):≤ 0.0200 %
- Related substances / HPLC (EP) (4-Hydroxybenzoic acid):≤ 0.1 %
- Related substances / HPLC (EP) (4-Hydroxyisophthalic acid):≤ 0.05 %
- Related substances / HPLC (EP) (Phenol):≤ 0.02 %
- Related substances / HPLC (EP) (Largest unspecified impurity):≤ 0.05 %
- Related substances / HPLC (EP) (Sum of all impurities):≤ 0.2 %
- Related substances / HPLC (USP) (4-Hydroxybenzoic acid):≤ 0.1 %
- Related substances / HPLC (USP) (4-Hydroxyisophthalic acid):≤ 0.05 %
- Related substances / HPLC (USP) (Phenol):≤ 0.02 %
- Related substances / HPLC (USP) (Largest unspecified impurity):≤ 0.05 %
- Related substances / HPLC (USP) (Sum of all impurities):≤ 0.2 %
- Residual solvents (ICH Q3C):excluded by manufacturing process
- Sulfated ash:≤ 0.05 %
- Loss on drying:≤ 0.5 %
Frequently Bought Together