Order Entry
Ireland
ContactUsLinkComponent
3-Nitrobenzoic acid for synthesis, Sigma-Aldrich®
Irritant
3-Nitrobenzoic acid for synthesis, Sigma-Aldrich®
Catalog # 8.00645.0250
Supplier:  Merck
CAS Number:  
Irritant
3-Nitrobenzoic acid for synthesis, Sigma-Aldrich®
Catalog # 8.00645.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Solubility:
    3 g/l (25 °C)
  • Melting Point:
    139 - 142 °C
  • Molar Mass:
    167.12 g/mol
  • Bulk Density:
    700 kg/m3
  • Flash Point:
    190 °C
  • Density:
    1.49 g/cm3 (20 °C)
  • pH:
    3 (5 g/l, H2O, 20 °C) (aqueous suspension)
  • Ignition Point:
    490 °C
  • Hill Formula:
    C7H5NO4
  • Chemical Formula:
    3-(NO2)C6H4COOH

Specification Test Results

About this item

 :
 :167.12 g/mol
Merck Index:12,06686
Boiling Pt:341 °C (1013 hPa)
Melting Pt:139…142 °C
Density:1,494 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:204-508-5
 :GHS07
CAS Number:121-92-6
Formula:C₇H₅NO₄
MDL Number:MFCD00007251