Order Entry
Ireland
ContactUsLinkComponent
3-Cyanobenzoic acid
Irritant
3-Cyanobenzoic acid
Catalog # MOLE60148293-25G
Supplier:  Molekula
CAS Number:  
Irritant
3-Cyanobenzoic acid
Catalog # MOLE60148293-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :147.13 g/mol
Melting Pt:219 °C
EINECS:217-511-1
 :GHS07
Storage Temperature:Ambient
CAS Number:1877-72-1
Formula:C₈H₅NO₂
MDL Number:MFCD00002486